Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D430042O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081022.1 | NP_001074491.1 | 15 | 2580 | ||
GenBank mRNA | AK052528.1 | - | 10 | 2087 | ||
BC157963.1 | - | 14 | 2512 | |||
BC158019.1 | - | 15 | 2568 | 9007325 |