Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033040.3 | NP_001028212.1 | 8 | 1269 | ||
GenBank mRNA | AK140121.1 | - | 8 | 1270 | ||
BC066021.1 | - | 8 | 850 | 5717506 |