Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024922.2 | NP_001020093.2 | 8 | 909 | ||
GenBank mRNA | BC085301.1 | - | 8 | 909 | ||
BC099554.1 | - | 8 | 916 | 30308124 |