Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colgalt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146211.3 | NP_666323.2 | 4 | 751 | ||
GenBank mRNA | AK029433.1 | - | 4 | 605 | ||
AK035788.1 | - | 4 | 629 | |||
AK036165.1 | - | 4 | 752 | |||
AK076103.1 | - | 4 | 607 | |||
AK135643.1 | - | 1 | 470 | |||
AK154775.1 | - | 4 | 606 | |||
BC018287.1 | - | 4 | 516 | |||
BC032165.1 | - | 4 | 783 | 5353334 | ||
BC034208.1 | - | 1 | 69 | |||
BC056951.1 | - | 4 | 642 | 5695490 |