Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205226.1 | NP_001192155.1 | 28, 29 | 4100 | ||
NM_153164.3 | NP_694804.3 | 28, 29 | 4106 | |||
NM_178078.2 | NP_835179.1 | 28, 29 | 4106 | |||
GenBank mRNA | AK034776.1 | - | 15, 16 | 2223 | ||
AK129258.1 | - | 1066 | ||||
BC018281.1 | - | 1, 2 | 77 | |||
BC035210.1 | - | 13, 14 | 2001 | |||
BC058249.1 | - | 28, 29 | 4056 | |||
BC157948.1 | - | 28, 29 | 4019 | 9088123 | ||
BC158073.1 | - | 28, 29 | 4025 | 9007370 | ||
BC172105.1 | - | 28, 29 | 4025 | 9054391 |