Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163756.1 | NP_001157228.1 | 11 | 1513 | ||
NM_172759.3 | NP_766347.1 | 11 | 1513 | |||
GenBank mRNA | AK077248.1 | - | 11 | 1504 | ||
BC022148.1 | - | 7 | 826 | |||
BC055062.1 | - | 11 | 1482 | 6528966 |