Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146214.3 | NP_666326.1 | 4 | 485 | ||
GenBank mRNA | AK090244.1 | - | 4 | 483 | ||
AK149383.1 | - | 4 | 483 | |||
AK149526.1 | - | 4 | 483 | |||
BC023949.1 | - | 4 | 469 | 5310059 | ||
BC024120.1 | - | 4 | 460 | 5097591 | ||
BC024264.1 | - | 3 | 408 | 5098682 | ||
BC025934.1 | - | 4 | 420 | 5097113 | ||
BC028821.1 | - | 4 | 460 | 5097589 | ||
BC030728.1 | - | 4 | 464 | 5099036 | ||
BC030729.1 | - | 4 | 458 | 5099570 | ||
BC037526.1 | - | 4 | 440 | 5097795 |