Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190800.1 | NP_001177729.1 | 1, 2 | 179 | ||
NM_172284.5 | NP_758488.1 | 1, 2 | 179 | |||
GenBank mRNA | AK013362.1 | - | 1, 2 | 174 | ||
AK147499.1 | - | 1, 2 | 174 | |||
BC025594.1 | - | 1, 2 | 139 | 5342859 |