Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145606.3 | NP_663581.1 | 6 | 570 | ||
GenBank mRNA | AK046416.1 | - | 6 | 543 | ||
AK159830.1 | - | 6 | 543 | |||
BC010524.1 | - | 6 | 520 | 3500937 | ||
BC023807.1 | - | 6 | 533 | 5354266 | ||
BC036138.1 | - | 6 | 522 | 4972627 | ||
BC036152.1 | - | 6 | 524 | 4977509 |