Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172290.3 | NP_758494.2 | 3 | 587 | ||
GenBank mRNA | AF282980.1 | - | 2 | 384 | ||
AK042854.1 | - | 3 | 589 | |||
AK045973.1 | - | 3 | 587 | |||
AK046377.1 | - | 3 | 587 | |||
AK138263.1 | - | 3 | 575 | |||
AK140516.1 | - | 3 | 589 | |||
AK158752.1 | - | 2 | 606 | |||
BC023307.1 | - | 2 | 427 | 4480983 |