Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gldn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177350.5 | NP_796324.1 | 9 | 1145 | ||
GenBank mRNA | AF548022.1 | - | 9 | 801 | ||
AK031523.1 | - | 9 | 1146 | |||
BC125025.1 | - | 9 | 1125 | 40058551 | ||
BC125026.1 | - | 9 | 1125 | 40058552 |