Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam214a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113283.1 | NP_001106754.1 | 6 | 1693 | ||
NM_153584.2 | NP_705812.2 | 5 | 1516 | |||
GenBank mRNA | AK173161.1 | - | 6 | 1564 | ||
BC031353.1 | - | 5 | 1516 | 5363230 | ||
BC060215.1 | - | 6 | 1691 |