Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430571L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175486.3 | NP_780695.1 | 6 | 859 | ||
GenBank mRNA | AK032358.1 | - | 6 | 860 | ||
AK032501.1 | - | 6 | 866 | |||
AK043338.1 | - | 6 | 899 | |||
AK085251.1 | - | 6 | 890 | |||
BC115775.1 | - | 6 | 771 | 40107115 | ||
BC116708.1 | - | 6 | 771 | 40107114 |