Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167775.1 | NP_001161247.1 | 9 | 1591 | ||
NM_001167776.1 | NP_001161248.1 | 8 | 1073 | |||
NM_145627.2 | NP_663602.1 | 9 | 1304 | |||
GenBank mRNA | AK041871.1 | - | 8 | 1048 | ||
AK089105.1 | - | 8 | 884 | |||
AK122208.1 | - | 7 | 2833 | |||
AK145991.1 | - | 9 | 1304 | |||
AK153736.1 | - | 9 | 1592 | |||
AK167666.1 | - | 8 | 1051 | |||
BC004674.1 | - | 8 | 989 | 3500403 | ||
BC093519.1 | - | 8 | 1005 |