Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146234.3 | NP_666346.1 | 4 | 399 | ||
GenBank mRNA | AK036253.1 | - | 4 | 400 | ||
AK043524.1 | - | 4 | 413 | |||
AK049689.1 | - | 4 | 374 | |||
AK163269.1 | - | 4 | 551 | |||
BC032271.1 | - | 4 | 425 | 4954726 | ||
EU069461.1 | - | 4 | 358 |