Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC023829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033328.2 | NP_001028500.1 | 4 | 529 | ||
GenBank mRNA | AK158294.1 | - | 4 | 529 | ||
BC138670.1 | - | 5 | 609 | 8861691 | ||
BC145368.1 | - | 5 | 609 | 9053926 |