Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011603.5 | NP_035733.1 | 3, 4 | 594 | ||
GenBank mRNA | AB017697.1 | - | 3, 4 | 491 | ||
AK005604.1 | - | 3, 4 | 414 | |||
AK049975.1 | - | 3, 4 | 466 | |||
AK078547.1 | - | 3, 4 | 526 | |||
BC033926.1 | - | 3, 4 | 435 | 1382180 |