Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153393.2 | NP_700442.2 | 4, 5 | 597 | ||
GenBank mRNA | AF410792.1 | - | 4, 5 | 597 |