Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sowaha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183173.2 | NP_898996.2 | 1 | 1894 | ||
GenBank mRNA | AK043538.1 | - | 1 | 1781 | ||
AY496686.1 | - | 1 | 363 | |||
BC117862.1 | - | 2 | 572 | |||
BC117863.1 | - | 1 | 1232 | 40095300 |