Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159519.1 | NP_001152991.1 | 10 | 1231 | ||
GenBank mRNA | AK139115.1 | - | 10 | 1223 | ||
BC114997.1 | - | 9 | 1165 | 30110327 |