Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011790.4 | NP_035920.1 | 6, 7 | 838 | ||
GenBank mRNA | AF124664.1 | - | 5, 6 | 665 | ||
AJ130975.1 | - | 5, 6 | 697 | |||
AK011094.1 | - | 6, 7 | 842 | |||
AK045012.1 | - | 821 | ||||
AK149936.1 | - | 6, 7 | 818 | |||
AK167104.1 | - | 6, 7 | 795 | |||
AK168055.1 | - | 6, 7 | 843 | |||
BC051998.1 | - | 6, 7 | 838 | 6400287 | ||
BC052422.1 | - | 6, 7 | 838 | 6400287 |