Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178714.4 | NP_848829.2 | 4 | 2102 | ||
GenBank mRNA | AK034245.1 | - | 4 | 2103 | ||
AK048443.1 | - | 4 | 2188 | |||
AK083920.1 | - | 4 | 3297 | |||
AK220546.1 | - | 4 | 2786 | |||
BC052038.1 | - | 4 | 1932 | 5708346 |