Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038231.2 | NP_001033320.1 | 3 | 351 | ||
NM_001286608.1 | NP_001273537.1 | 3 | 693 | |||
NM_011793.3 | NP_035923.1 | 3 | 559 | |||
GenBank mRNA | AB025349.1 | - | 3 | 291 | ||
AF037080.1 | - | 2 | 234 | |||
AK003025.1 | - | 3 | 335 | |||
AK004212.1 | - | 3 | 318 | |||
AK010257.1 | - | 3 | 337 | |||
AK011079.1 | - | 3 | 340 | |||
AK011100.1 | - | 3 | 338 | |||
AK012588.1 | - | 3 | 321 | |||
AK088896.1 | - | 3 | 315 | |||
AK168039.1 | - | 3 | 340 | |||
BC003709.1 | - | 3 | 328 | 3599828 | ||
CK780850.1 | - | 3 | 323 |