Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027185.3 | NP_081461.2 | 3 | 427 | ||
GenBank mRNA | AF329497.1 | - | 3 | 362 | ||
AK038050.1 | - | 3 | 398 | |||
AK088460.1 | - | 3 | 376 | |||
AY241695.1 | - | 3 | 362 | |||
BC120500.1 | - | 3 | 427 | 40129423 | ||
BC131953.1 | - | 3 | 427 | 40130230 | ||
X96705.1 | - | 3 | 319 |