Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata31d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167593.1 | NP_001161065.1 | 4 | 1442 | ||
GenBank mRNA | AK133300.1 | - | 4 | 1444 |