Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011819.2 | NP_035949.2 | 2 | 767 | ||
GenBank mRNA | AF159571.1 | - | 2 | 1238 | ||
AK144199.1 | - | 3 | 1066 | |||
AK159242.1 | - | 2 | 768 | |||
AK160927.1 | - | 2 | 680 | |||
BC067248.1 | - | 2 | 756 | 820434 |