Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011826.4 | NP_035956.1 | 3 | 549 | ||
GenBank mRNA | AF023482.1 | - | 3 | 521 | ||
AF023483.1 | - | 2 | 684 | |||
AK002256.1 | - | 3 | 500 | |||
AK010633.1 | - | 3 | 533 | |||
AK088746.1 | - | 3 | 531 | |||
BC006688.1 | - | 3 | 478 | 3582652 | ||
BC098225.1 | - | 3 | 503 | 1494110 | ||
BC147481.1 | - | 3 | 480 | 9056226 |