Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011827.3 | NP_035957.2 | 2, 3 | 176 | ||
GenBank mRNA | AF072846.1 | - | 2, 3 | 68 | ||
AF172930.1 | - | 2, 3 | 176 | |||
AK008005.1 | - | 2, 3 | 175 | |||
BC145545.1 | - | 2, 3 | 167 | 9054109 | ||
BC145824.1 | - | 2, 3 | 167 | 40131127 | ||
BC145826.1 | - | 2, 3 | 167 | 40131129 |