Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011832.2 | NP_035962.2 | 6 | 1820 | ||
GenBank mRNA | AB007135.1 | - | 6 | 1709 | ||
AK081512.1 | - | 6 | 1807 | |||
AK142578.1 | - | 6 | 1821 | |||
BC137855.1 | - | 6 | 1677 | 8860874 |