Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178719.5 | NP_848834.2 | 3 | 584 | ||
GenBank mRNA | AK038466.1 | - | 3 | 557 | ||
AK041184.1 | - | 3 | 563 | |||
AK044773.1 | - | 4 | 1017 | |||
AK050546.1 | - | 3 | 545 | |||
AK080375.1 | - | 3 | 530 | |||
AK163389.1 | - | 4 | 685 | |||
BC094597.1 | - | 3 | 516 |