Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177356.3 | NP_796330.2 | 5 | 1186 | ||
GenBank mRNA | AJ510130.1 | - | 5 | 1105 | ||
AK028039.1 | - | 5 | 1138 | |||
AK033379.1 | - | 5 | 1139 |