Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnldc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034866.1 | NP_001030038.1 | 12, 13 | 1089 | ||
GenBank mRNA | BC158033.1 | - | 12, 13 | 1089 | 9007336 | |
BC172002.1 | - | 12, 13 | 1092 |