Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011879.2 | NP_036009.1 | 1, 2 | 134 | ||
GenBank mRNA | AJ006130.1 | - | 1, 2 | 17 | ||
AK029644.1 | - | 1, 2 | 88 | |||
AK050625.1 | - | 1, 2 | 92 | |||
AK082356.1 | - | 1, 2 | 66 | |||
AK087976.1 | - | 1, 2 | 80 | |||
AK088908.1 | - | 1, 2 | 69 | |||
AK163766.1 | - | 1, 2 | 88 | |||
AK167238.1 | - | 1, 2 | 99 | |||
BC006706.1 | - | 1, 2 | 112 | |||
BC014739.1 | - | 1, 2 | 65 | 4920184 | ||
BC026569.1 | - | 1, 2 | 65 | 4982612 | ||
BC029148.1 | - | 1, 2 | 107 | |||
BC029671.1 | - | 1, 2 | 77 | 5364953 | ||
BC029694.1 | - | 1, 2 | 74 | 5369477 | ||
BC033356.1 | - | 1, 2 | 61 | 5366230 | ||
BG084120.2 | - | 1, 2 | 134 |