Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243307.1 | NP_001230236.1 | 2 | 426 | ||
NM_198616.4 | NP_941018.1 | 1 | 581 | |||
GenBank mRNA | AK140354.1 | - | 1 | 915 | ||
BC016659.1 | - | 2 | 361 | |||
BC058411.1 | - | 1 | 567 | 5710206 | ||
BC094592.1 | - | 2 | 367 | |||
BQ445696.1 | - | 1 | 567 |