Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011890.4 | NP_036020.1 | 5 | 713 | ||
GenBank mRNA | AB024921.1 | - | 5 | 727 | ||
AK014381.2 | - | 5 | 727 | |||
AK017781.1 | - | 5 | 717 | |||
AK132306.1 | - | 5 | 772 | |||
AK169403.1 | - | 5 | 714 | |||
BC052349.1 | - | 5 | 703 | 6744003 |