Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172840.2 | NP_766428.2 | 9 | 1030 | ||
GenBank mRNA | AJ536329.1 | - | 9 | 1030 | ||
AK029297.1 | - | 9 | 977 | |||
AK133486.1 | - | 9 | 937 | |||
AK145058.1 | - | 9 | 1200 | |||
AK160971.1 | - | 9 | 1036 | |||
AK166339.1 | - | 9 | 1108 | |||
BC115868.1 | - | 8 | 932 | 40081354 | ||
BC116636.1 | - | 9 | 980 | 40081359 |