Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033344.3 | NP_001028516.2 | 6 | 2866 | ||
NM_001160049.1 | NP_001153521.1 | 6 | 2862 | |||
GenBank mRNA | AK048603.1 | - | 6 | 2889 | ||
AK142263.1 | - | 6 | 2867 | |||
BC059034.1 | - | 2 | 2215 | |||
BC117936.1 | - | 6 | 2843 | 40097789 |