Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011908.2 | NP_036038.1 | 3 | 980 | ||
GenBank mRNA | AF044222.1 | - | 3 | 922 | ||
AF044223.1 | - | 3 | 241 | |||
AK134598.1 | - | 3 | 348 | |||
AK145952.1 | - | 3 | 659 | |||
BC024507.1 | - | 5 | 830 | 4983604 | ||
BC025595.1 | - | 3 | 635 | 5342735 | ||
BC043729.1 | - | 5 | 830 | 4983604 |