Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163566.1 | NP_001157038.1 | 6, 7 | 1166 | ||
GenBank mRNA | BC043114.1 | - | 5, 6 | 854 |