Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290986.1 | NP_001277915.1 | 10, 11 | 1464 | ||
NM_001290987.1 | NP_001277916.1 | 9, 10 | 1349 | |||
NM_030234.2 | NP_084510.2 | 10, 11 | 1479 | |||
GenBank mRNA | AK031592.1 | - | 10, 11 | 1405 | ||
AK162181.1 | - | 10, 11 | 1432 | |||
BC094676.1 | - | 9, 10 | 1115 | 30615673 | ||
BC117821.1 | - | 9, 10 | 1288 | |||
BC117822.1 | - | 10, 11 | 1418 | 40092839 |