Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291027.1 | NP_001277956.1 | 4 | 1221 | ||
NM_001291028.1 | NP_001277957.1 | 4 | 879 | |||
NM_197945.4 | NP_922936.3 | 4 | 1221 | |||
GenBank mRNA | AK161711.1 | - | 4 | 1175 | ||
BC049892.1 | - | 2 | 782 | |||
BC058280.1 | - | 3 | 803 | 6529102 |