Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183181.2 | NP_899004.1 | 1 | 832 | ||
GenBank mRNA | AK028091.1 | - | 1 | 834 | ||
BC050777.1 | - | 1 | 828 | 6774737 |