Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287224.1 | NP_001274153.1 | 3, 4 | 885 | ||
GenBank mRNA | KF954709.1 | - | 3, 4 | 885 |