Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033351.2 | NP_001028523.1 | 5 | 3287 | ||
NM_001276355.1 | NP_001263284.1 | 5 | 3287 | |||
GenBank mRNA | AK046827.1 | - | 1 | 107 | ||
AK138366.1 | - | 5 | 3288 | |||
AK173314.1 | - | 5 | 2288 |