Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158868.1 | - | 10 | 3131 | ||
AY243848.2 | - | 8 | 2511 | |||
BC057615.1 | - | 10 | 2740 | 6401651 | ||
BC058433.1 | - | 8 | 1938 | |||
BC098311.1 | - | 3273 |