Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C130060K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175524.4 | NP_780733.2 | 4 | 759 | ||
GenBank mRNA | AK048439.1 | - | 4 | 669 | ||
AK051723.1 | - | 4 | 693 | |||
BC119578.1 | - | 4 | 660 | 40058179 | ||
BC119579.1 | - | 4 | 660 | 40058180 |
Set Membership: |
G-protein coupled receptor |