Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159507.1 | NP_001152979.1 | 21 | 2695 | ||
GenBank mRNA | BC099701.1 | - | 12 | 1584 |