Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146259.3 | NP_666371.1 | 3 | 2618 | ||
GenBank mRNA | AK047593.1 | - | 4 | 2658 | ||
AK053438.1 | - | 3 | 2619 | |||
AK152456.1 | - | 3 | 1911 | |||
AK169556.1 | - | 3 | 2624 | |||
BC028251.1 | - | 3 | 2981 | |||
BC030314.1 | - | 3 | 2234 | 5371636 |