Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146259.3 | NP_666371.1 | 2 | 852 | ||
GenBank mRNA | AK047593.1 | - | 3 | 892 | ||
AK053438.1 | - | 2 | 853 | |||
AK081436.1 | - | 2 | 848 | |||
AK152456.1 | - | 2 | 145 | |||
AK169556.1 | - | 2 | 858 | |||
BC028251.1 | - | 2 | 1215 | |||
BC030314.1 | - | 2 | 468 | 5371636 |