Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5830411N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128145.1 | NP_001121617.1 | 2 | 219 | ||
NM_001128146.1 | NP_001121618.1 | 2 | 222 | |||
NM_175533.3 | NP_780742.1 | 2 | 299 | |||
GenBank mRNA | AK030813.1 | - | 2 | 317 | ||
AK040624.1 | - | 2 | 300 | |||
AK220311.1 | - | 2 | 270 | |||
BC139294.1 | - | 2 | 267 | |||
EF624462.1 | - | 2 | 222 | |||
EF624463.1 | - | 2 | 222 |